Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric acid-[D₂] 96% (99.5% D) (w/w) in D₂O for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Sulphuric acid-[²H₂] 98%

87857.18 87857-18
AA87857-18EA 241.99 USD InStock
CAAA87857-18 AA87857-18
Sulfuric acid-[D₂] 96% (99.5% D) (w/w) in D₂O for NMR spectroscopy
Sulfuric acid-[D₂]


Formula: D₂SO₄
MW: 100.09 g/mol
Boiling Pt: 290 °C
Density: 1.86
Storage Temperature: Ambient
MDL Number: MFCD00003542
CAS Number: 13813-19-9
EINECS: 237-464-0
UN: 1830
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty