Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Decaborane 98%

Supplier: Thermo Scientific Chemicals
Danger

087892-03 087892-06 087892-14
AA87892-03EA 82.09 USD InStock
AA87892-03 AA87892-14 AA87892-06
Decaborane 98%
Decaborane
Slightly soluble in cold water. Hydrolyzes in hot water. Soluble in ethyl acetate, 1-Bromopropane, ethyl silicate, benzene, alcohol, and carbon tetrachloride

In rocket propellants, and as a catalyst in olefin polymerization


Formula: B₁₀H₁₄
Boiling Pt: 213 °C
Melting Pt: 99…103 °C
Density: 0.940
Storage Temperature: Ambient
MDL Number: MFCD00011314
CAS Number: 17702-41-9
EINECS: 241-711-8
UN: 1868
ADR: 4.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty