Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitric acid 65 - 70% ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Aqua fortisAzotic acidHydrogen nitrate

87920-AP 087920-AE 087920-AP 87920-AE
AA87920-AEEA 90.74 USD InStock
AA87920-AE CAAA87920-AP AA87920-AP CAAA87920-AE
Nitric acid 65 - 70% ≥99.999% (metals basis)
Nitric acid
In the manufacture of fertilizers and explosives, in organic synthesis, in metallurgy, in photoengraving, in etching steel, in rubber chemicals, and in reprocessing nuclear fuels


Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 121 °C
Melting Pt: –42 °C
Density: 1.41
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II
Merck Index: 13,06608
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty