Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorosulfuric acid ∼99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Chlorosulphonic acid

87977-30 087977-A1 087977-30 087977-D5 87977-D5 87977-A1
AA87977-A1EA 506.26 USD InStock
AA87977-A1 CAAA87977-D5 CAAA87977-A1 AA87977-30 AA87977-D5 CAAA87977-30
Chlorosulfuric acid ∼99%
Chlorosulphuric acid


Formula: HClO₃S
MW: 116.52 g/mol
Boiling Pt: 158 °C
Melting Pt: –80 °C
Density: 1,753
Storage Temperature: Ambient
MDL Number: MFCD00011523
CAS Number: 7790-94-5
EINECS: 232-234-6
UN: 1754
ADR: 8,I
Merck Index: 12,02218
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty