Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zirconium, AAS Standard Solution, Specpure®, Zr 1000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific
Specpure®
88119-AP 088119-AP
AA88119-APEA 81.14 USD InStock
AA88119-AP CAAA88119-AP
Zirconium, AAS Standard Solution, Specpure®, Zr 1000 µg/ml, Thermo Scientific
Standards Cationic Standards Zirconium Standards
Molecular Formula: ZrCl2O in 5% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011303

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty