Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Strontium oxide ≥99.5% (metals basis) SrO ≈97%, Powder -100 Mesh

Supplier: Thermo Scientific Chemicals
Danger

088220-14 88220-06 88220-22 088220-22 88220-14 088220-06
AA88220-22EA 429.99 USD InStock
AA88220-22 CAAA88220-14 CAAA88220-22 CAAA88220-06 AA88220-06 AA88220-14
Strontium oxide ≥99.5% (metals basis) SrO ≈97%, Powder -100 Mesh
Strontium oxide
Strontium oxide was used to make television picture tube glass. It finds application in glass, optic and ceramic industry. It is an active component of paint dryers. It is also used to prepare pure strontium up on heating with aluminum under vacuum.

Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with water.


Formula: SrO
Boiling Pt: ∼3000 °C
Melting Pt: 2430 °C
Density: 4,700
Storage Temperature: Ambient
MDL Number: MFCD00049558
CAS Number: 1314-11-0
EINECS: 215-219-9
UN: 3262
ADR: 8,II
Merck Index: 12,09009
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty