088220-14
88220-06
88220-22
088220-22
88220-14
088220-06
AA88220-22EA
429.99
USD
InStock
AA88220-22
CAAA88220-14
CAAA88220-22
CAAA88220-06
AA88220-06
AA88220-14
Strontium oxide ≥99.5% (metals basis) SrO ≈97%, Powder -100 Mesh
Strontium oxide
Strontium oxide was used to make television picture tube glass. It finds application in glass, optic and ceramic industry. It is an active component of paint dryers. It is also used to prepare pure strontium up on heating with aluminum under vacuum.
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with water.