Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium cyclopentadienide 2-3 M in THF

Supplier: Thermo Scientific Chemicals
Danger

088300-JQ 88300-JQ 88300-JT 088300-JT
AA88300-JTEA 425.46 USD InStock
AA88300-JT CAAA88300-JT AA88300-JQ CAAA88300-JQ
Sodium cyclopentadienide 2-3 M in THF
Sodium cyclopentadienide
Reagent for the preparation of metallocenes


Formula: C₅H₅Na
Density: 0,938
Flash Pt: –17 °C (1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012161
CAS Number: 4984-82-1
EINECS: 225-636-8
UN: 3399
ADR: 4.3,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty