Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hydroxide concentrate for dilution to 0.5 N in aqueous solution, Acculute™ Standard Volumetric Solution

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Potassium hydrateCaustic potash

088602-LQ 088602-LS 88602-LQ 88602-LS
AA88602-LSEA 349.99 USD InStock
AA88602-LS AA88602-LQ CAAA88602-LQ CAAA88602-LS
Potassium hydroxide concentrate for dilution to 0.5 N in aqueous solution, Acculute™ Standard Volumetric Solution
Potassium hydroxide


Formula: KOH
MW: 56.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty