Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide concentrate for dilution to 0.5 N in aqueous solution, Acculute™ Standard Volumetric Solution

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

088620-LS 88620-LS
AA88620-LSEA 160.99 USD InStock
AA88620-LS CAAA88620-LS
Sodium hydroxide concentrate for dilution to 0.5 N in aqueous solution, Acculute™ Standard Volumetric Solution
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: ∼105 °C (1013 hPa)
Density: 1.327 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty