Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EDTA ferric monosodium salt

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Iron (III) sodium ethylenediaminetetraacetate Edetic acid iron (III) sodium salt Ethylenediaminetetra-acetic acid iron (III) sodium salt Ethylenedinitrilotetra-acetic acid iron (III) sodium salt Edetic acid iron (III) monosodium salt Ethylenediaminetetra-acetic acid iron (III) monosodium salt Ethylenedinitrilotetra-acetic acid iron (III) monosodium salt Iron (III) monosodium ethylenediaminetetraacetate Sodium feredetate

088995-36 88995-A3 088995-A3 088995-22 88995-36 88995-22
AA88995-22EA 48.55 USD InStock
CAAA88995-22 CAAA88995-A3 AA88995-22 CAAA88995-36 AA88995-A3 AA88995-36
EDTA ferric monosodium salt
EDTA ferric monosodium salt


Formula: C₁₀H₁₂FeN₂NaO₈
MW: 367.05 g/mol
Melting Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00078215
CAS Number: 15708-41-5
EINECS: 239-802-2
Merck Index: 12,04076
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty