Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Borane-d3 1 M in THF

Supplier: Thermo Scientific Chemicals
Danger

089121-AK 089121-AD
AA89121-AKEA 2308.99 USD InStock
AA89121-AK AA89121-AD
Borane-d3 1 M in THF
Borane-d3


Formula: BH₃
Density: 0,876
Flash Pt: –21 °C (–6 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD08705283
CAS Number: 13763-62-7
UN: 3148
ADR: 4.3,I
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty