Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Borane dimethylamine complex ≥97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Boron hydride dimethylamineBorane, dimethylamine complex

089149-14 089149-22 89149-22 89149-14
AA89149-14EA 81.86 USD InStock
AA89149-14 CAAA89149-14 AA89149-22 CAAA89149-22
Borane dimethylamine complex ≥97%
Borane dimethylamine complex


Formula: C₂H₁₀BN
MW: 58.92 g/mol
Boiling Pt: 59…65 °C (1…2 mmHg)
Melting Pt: 36 °C
Density: 0,69
Storage Temperature: Ambient
MDL Number: MFCD00051068
CAS Number: 74-94-2
EINECS: 200-823-7
UN: 2925
ADR: 4.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty