Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium cyanamide, tech.

Supplier: Thermo Scientific Chemicals
Danger

89462-22 89462-22 89462-36 089462-36
AA89462-36EA 152.99 USD InStock
AA89462-36 AA89462-22 CAAA89462-36 CAAA89462-22
Calcium cyanamide, tech.
Calcium cyanamide


Formula: CCaN₂
Melting Pt: 1340 °C
Density: 2.29
Storage Temperature: Ambient
MDL Number: MFCD00064894
CAS Number: 156-62-7
EINECS: 205-861-8
UN: 1403
ADR: 4.3,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty