Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rhenium disulfide 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Rhenium (IV) sulphide

089482-04 089482-09 89482-04 89482-09
AA89482-04EA 397.99 USD InStock
CAAA89482-04 CAAA89482-09 AA89482-04 AA89482-09
Rhenium disulfide 99%
Rhenium disulphide


Formula: ReS₂
MW: 250.34 g/mol
Density: 7.506
Storage Temperature: Ambient
MDL Number: MFCD00016307
CAS Number: 12038-63-0
EINECS: 234-878-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty