Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diphosphorus pentoxide ≥99.99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Phosphorus pentoxide Phosphoric anhydride

089966-22 89966-09 089966-36 089966-09 89966-36 89966-22
AA89966-22EA 127.29 USD InStock
CAAA89966-22 AA89966-22 CAAA89966-36 AA89966-36 CAAA89966-09 AA89966-09
Diphosphorus pentoxide ≥99.99%
di-Phosphorus pentoxide


Formula: P₂O₅
MW: 141.94 g/mol
Boiling Pt: 360 °C (subl.)
Melting Pt: 340 °C
Density: 2.39
Storage Temperature: Ambient
MDL Number: MFCD00011440
CAS Number: 1314-56-3
EINECS: 215-236-1
UN: 1807
ADR: 8,II
Merck Index: 13,07441
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty