Would you like to log in again?
Glove Bag, Polyethylene, Thermo Scientific
Supplier: Thermo Scientific
092727-LL 091717-LK 093727-LL 092717-LL 092727-LK 093737-LL 093727-LK 093737-LK 092717-LK 091717-LL
AA93727-LLEA 568.78 USD InStock
AA93727-LL AA93727-LK AAA92717-LK AA93737-LL AAA93737-LK AA91717-LL AAA91717-LK AAA92727-LK AA92717-LL AA92727-LL
Glove Bag, Polyethylene, Thermo Scientific
Glove Boxes and Glove Bags
Polyethylene glove bag is used for laboratory use and field sampling
Suitable for handling the air and moisture sensitive, high purity chemicals and materials.
Delivery information: Bag includes closure, tubing coupler and cotton gloves.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty