Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Methylethylenediamine 95% stabilized

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-AminoethylmethylamineN1-Methylethane-1,2-diamine

A12165-22 A12165-22 A12165-14 A12165-06 A12165-14 A12165-06
AAA-A12165-06EA 69.39 USD InStock
AAA-A12165-06 AAA-A12165-22 CAAAA12165-14 CAAAA12165-06 CAAAA12165-22 AAA-A12165-14
N-Methylethylenediamine 95% stabilized
N-Methylethylenediamine


Formula: C₃H₁₀N₂
MW: 74.13 g/mol
Boiling Pt: 114…116 °C
Density: 0.851
Flash Pt: 41 °C(105 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008165
CAS Number: 109-81-9
EINECS: 203-707-4
UN: 2734
ADR: "8",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty