Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Chloronaphthalene tech. 85% remainder 2-chloronaphthalene, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Triisodecyl phosphiteα-Chloronaphthalene

A15029-30 A15029-18 A15029-30 A15029-0B
AAA-A15029-0BEA 313.99 USD InStock
AAA-A15029-0B AAA-A15029-18 AAAA15029-30 CAAAA15029-30
1-Chloronaphthalene tech. 85% remainder 2-chloronaphthalene, Technical Grade
1-Chloronaphthalene


Formula: C₁₀H₇Cl
MW: 162.62 g/mol
Boiling Pt: 259 °C
Melting Pt: ca …8 °C
Density: 1.194
Flash Pt: 121 °C(249 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003874
CAS Number: 90-13-1
EINECS: 201-967-3
UN: 3082
ADR: "9",III
Merck Index: 13,02168
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty