Synonyms:
Sodium 4-(3,5-dichloro-4-oxocyclohexa-2,5-dienylideneamino)phenoxide hydrate Tillmans' reagent hydrate 2,6-Dichloroindophenol sodium hydrate
A10107-14
A10107-06
A10107-22
A10107-22
A10107-14
A10107-06
AAA10107-22EA
624.99
USD
InStock
CAAAA10107-06
AAA10107-22
CAAAA10107-14
CAAAA10107-22
AAA10107-06
AAA10107-14
2,6-Dichlorophenolindophenol sodium salt hydrate
2,6-Dichlorophenolindophenol sodium salt hydrate
Suitable for ascorbic acid determination
2,6-Dichloroindophenol sodium salt hydrate is suitable for vitamin C determination. It acts as a redox dye to investigate the efficiency of gold nanoparticles (Au-NP) for the enzymatic activity of glucose oxidase.
It is also used to compose UVB specific dosimeter and to measure the rate of photosynthesis. It serves as a good reducing agent.