Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloropyrazine 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Chloropyrazine

A10108-14 A10108-22 A10108-06 A10108-14 A10108-22 A10108-06
AAA10108-06EA 53.89 USD InStock
CAAAA10108-06 AAA10108-06 CAAAA10108-14 CAAAA10108-22 AAA10108-22 AAA10108-14
2-Chloropyrazine 98%
2-Chloropyrazine


Formula: C₄H₃ClN₂
MW: 114.53 g/mol
Boiling Pt: 153…154 °C
Density: 1.284
Flash Pt: 57 °C(134 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006124
CAS Number: 14508-49-7
EINECS: 238-517-0
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty