Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Toluene-α-sulfonyl chloride 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Phenylmethanesulphonyl chloride α-Toluenesulphonyl chloride Phenylmethylsulphonyl chloride

A10124-06 A10124-22 A10124-22 A10124-06 A10124-14 A10124-14
AAA10124-06EA 40.78 USD InStock
AAA10124-06 CAAAA10124-22 CAAAA10124-14 AAA10124-22 CAAAA10124-06 AAA10124-14
Toluene-α-sulfonyl chloride 99%
Toluene-α-sulphonyl chloride


Formula: C₇H₇ClO₂S
MW: 190.65 g/mol
Boiling Pt: 120 °C/10 mm
Melting Pt: 89…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00007455
CAS Number: 1939-99-7
EINECS: 217-717-1
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty