Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methyl-3-(2-furyl)propenal 97%

Supplier: Thermo Scientific Chemicals
Warning

A10125-09 A10125-09 A10125-30 A10125-18 A10125-30 A10125-18
AAA10125-09EA 51.59 USD InStock
AAA10125-09 CAAAA10125-30 AAA10125-18 AAA10125-30 CAAAA10125-18 CAAAA10125-09
2-Methyl-3-(2-furyl)propenal 97%
2-Methyl-3-(2-furyl)propenal


Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 280…281 °C
Density: 1.097
Flash Pt: >100 °C(212 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00063240
CAS Number: 874-66-8
EINECS: 212-866-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty