Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,4-Dithioerythritol 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (R*,S*)-1,4-Dimercaptobutane-2,3-diol erythro-1,4-Dimercaptobutane-2,3-diol Dithioerythritol DTE

A10138-14 A10138-06 A10138-06 A10138-14
AAA10138-14EA 665.02 USD InStock
CAAAA10138-06 AAA10138-14 CAAAA10138-14 AAA10138-06
1,4-Dithioerythritol 99%
1,4-Dithioerythritol
Reagent for maintaining thiols in reduced state.


Formula: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Melting Pt: 82…84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063750
CAS Number: 6892-68-8
EINECS: 229-998-8
UN: 3335
ADR: "9",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty