Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pentyl benzoate 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: Benzoic acid pentyl ester Benzoic acid amyl ester Amyl benzoate

A10177-22 A10177-22 A10177-36 A10177-36
AAA10177-36EA 161.99 USD InStock
CAAAA10177-22 AAA10177-36 AAA10177-22 CAAAA10177-36
Pentyl benzoate 98+%
Pentyl benzoate


Formula: C₁₂H₁₆O₂
MW: 192.26 g/mol
Boiling Pt: 260 °C
Density: 0.99
Flash Pt: 130 °C(266 °F)
Storage Temperature: Ambient
MDL Number: MFCD00048843
CAS Number: 2049-96-9
EINECS: 218-077-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty