Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-Methyl-2-aminopropanoate hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Synonyms: H-Ala-OMe·HCl(S)-2-Aminopropionic Acid Methyl Ester Hydrochloride

A10178-14 A10178-06 A10178-22 A10178-06 A10178-22 A10178-14
AAA10178-22EA 373.28 USD InStock
CAAAA10178-14 AAA10178-22 CAAAA10178-06 AAA10178-14 AAA10178-06 CAAAA10178-22
(S)-Methyl-2-aminopropanoate hydrochloride 99%
(S)-Methyl-2-aminopropanoate hydrochloride


Formula: C4H10ClNO2
MW: 139.6 g/mol
Melting Pt: 106…110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063663
CAS Number: 2491-20-5
EINECS: 219-652-4
Order Now
Specification Test Results
Optical rotation +6.5° (c=10 in methanol)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty