Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Bis(trifluoromethyl)phenol 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: α,α,α,α',α',α'-Hexafluoro-3,5-xylenol

A10183-06 A10183-03 A10183-14 A10183-03 A10183-14 A10183-06
AAA10183-06EA 103.99 USD InStock
AAA10183-06 AAA10183-03 AAA10183-14 CAAAA10183-06 CAAAA10183-14 CAAAA10183-03
3,5-Bis(trifluoromethyl)phenol 97%
3,5-Bis(trifluoromethyl)phenol


Formula: C₈H₄F₆O
MW: 230.11 g/mol
Boiling Pt: 97…98 °C/50 mm
Melting Pt: 20…21 °C
Density: 1,511
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000386
CAS Number: 349-58-6
EINECS: 206-488-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty