Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Picolyl chloride hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-(Chloromethyl)pyridine hydrochloride

A10226-22 A10226-14 A10226-14 A10226-06 A10226-22 A10226-06
AAA10226-14EA 114.99 USD InStock
CAAAA10226-14 AAA10226-14 CAAAA10226-06 CAAAA10226-22 AAA10226-06 AAA10226-22
2-Picolyl chloride hydrochloride 98%
2-Picolyl chloride hydrochloride


Formula: C₆H₇Cl₂N
MW: 164.03 g/mol
Melting Pt: 120…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00012811
CAS Number: 6959-47-3
EINECS: 230-149-9
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty