Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Alanine 99%

Supplier: Thermo Scientific Chemicals
Synonyms: D(-)-Alanine (R)-(-)-Alanine Alanine α-Alanine H-D-Ala-OH D-α-Alanine

A10231-14 A10231-22 A10231-06 A10231-22 A10231-14 A10231-06
AAA10231-06EA 39.36 USD InStock
AAA10231-06 AAA10231-14 CAAAA10231-22 CAAAA10231-14 CAAAA10231-06 AAA10231-22
D-Alanine 99%
D-Alanine


Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: ca 295 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008077
CAS Number: 338-69-2
EINECS: 206-418-1
Merck Index: 13,00203
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty