Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Ethylbenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

A10235-30 A10235-18 A10235-09 A10235-18 A10235-30 A10235-09
AAA10235-30EA 618.99 USD InStock
AAA10235-30 CAAAA10235-30 AAA10235-18 AAA10235-09 CAAAA10235-18 CAAAA10235-09
4-Ethylbenzoic acid 99%
4-Ethylbenzoic acid


Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 270 °C (1013 hPa)
Melting Pt: 111…114 °C
Density: ∼0,9961 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002570
CAS Number: 619-64-7
EINECS: 210-605-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty