Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Atropine sulfate Monohydrate ≥98%

Supplier: Thermo Scientific Chemicals
Danger

A10236-18 A10236-18 A10236-09 A10236-09
AAA10236-09EA 68.66 USD InStock
AAA10236-09 CAAAA10236-09 AAA10236-18 CAAAA10236-18
Atropine sulfate Monohydrate ≥98%
Atropine sulfate Monohydrate
Antagonist at muscarinic receptors; also a bronchodilator


Formula: C₁₇H₂₃NO₃
Melting Pt: 187…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00074815
CAS Number: 5908-99-6
EINECS: 200-235-0
UN: 1544
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty