Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Polyethylene, powder 500 micron, low density

Supplier: Thermo Scientific Chemicals

A10239-0E A10239-22 A10239-36 A10239-36 A10239-22 A10239-0E
AAA10239-22EA 53.82 USD InStock
CAAAA10239-0E AAA10239-22 CAAAA10239-36 CAAAA10239-22 AAA10239-0E AAA10239-36
Polyethylene, powder 500 micron, low density
Polyethylene (CAS 9002-88-4)
for GLC


Formula: (CH₂CH₂)n
Density: 0.92
Storage Temperature: Ambient
MDL Number: MFCD00084423
CAS Number: 9002-88-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty