Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Oxoglutaric acid 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Ketoglutaric acid α-Ketoglutaric acid

A10256-36 A10256-22 A10256-14 A10256-14 A10256-36 A10256-22
AAA10256-22EA 75.71 USD InStock
CAAAA10256-22 CAAAA10256-14 CAAAA10256-36 AAA10256-22 AAA10256-36 AAA10256-14
2-Oxoglutaric acid 98%
2-Oxoglutaric acid


Formula: HOOCCH₂CH₂COCOOH
MW: 146.1 g/mol
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 113…117 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004165
CAS Number: 328-50-7
EINECS: 206-330-3
Merck Index: 13,05320
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty