Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Naphthoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: α-Naphthoic acid

A10273-22 A10273-14 A10273-22 A10273-14
AAA10273-22EA 85.73 USD InStock
AAA10273-22 CAAAA10273-22 CAAAA10273-14 AAA10273-14
1-Naphthoic acid 98%
1-Naphthoic acid


Formula: C₁₁H₈O₂
MW: 172.18 g/mol
Boiling Pt: >300 °C
Melting Pt: 160…162 °C
Density: 1,398
Flash Pt: 195 °C(383 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004007
CAS Number: 86-55-5
EINECS: 201-681-9
Merck Index: 12,06468
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty