Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chlorobenzenesulfonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3-Chlorobenzene-1-sulphonyl chloride

A10283-03 A10283-14 A10283-14 A10283-03 A10283-06 A10283-06
AAA10283-14EA 343.99 USD InStock
CAAAA10283-03 CAAAA10283-14 AAA10283-14 AAA10283-03 CAAAA10283-06 AAA10283-06
3-Chlorobenzenesulfonyl chloride 98%
3-Chlorobenzenesulphonyl chloride


Formula: C₆H₄Cl₂O₂S
MW: 211.07 g/mol
Boiling Pt: 102…104 °C/1 mm
Melting Pt: ca 21 °C
Density: 1.499
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00051697
CAS Number: 2888-06-4
EINECS: 220-753-0
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty