Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

o-Anisidine 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Methoxyaniline 2-Aminoanisole 2-Anisidine

A10288-0I A10288-0B A10288-30 A10288-0I A10288-0B A10288-30
AAA10288-0BEA 119.99 USD InStock
AAA10288-0B CAAAA10288-0I AAA10288-30 CAAAA10288-30 AAA10288-0I CAAAA10288-0B
o-Anisidine 99%
o-Anisidine


Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 224…225 °C
Melting Pt: 5…6 °C
Density: 1.092
Flash Pt: 98 °C(208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007688
CAS Number: 90-04-0
EINECS: 201-963-1
UN: 2431
ADR: "6.1",III
Merck Index: 13,00670
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty