Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Chlorosuccinimide 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1-Chloropyrrolidine-2,5-dione

A10310-18 A10310-0B A10310-30 A10310-30 A10310-18 A10310-0B
AAA10310-0BEA 162.99 USD InStock
AAA10310-0B AAA10310-30 CAAAA10310-18 CAAAA10310-30 AAA10310-18 CAAAA10310-0B
N-Chlorosuccinimide 98%
N-Chlorosuccinimide
Regioselective chlorination and oxidizing reagent


Formula: C₄H₄ClNO₂
MW: 133.53 g/mol
Boiling Pt: 216.5 °C (1013 hPa)
Melting Pt: 146…150 °C
Density: 1.65
Storage Temperature: Ambient
MDL Number: MFCD00005511
CAS Number: 128-09-6
EINECS: 204-878-8
UN: 3261
ADR: "8",III
Merck Index: 13,02183
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty