Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4,4'-Isopropylidenediphenol 97+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Bisphenol A 2,2-Bis(4-hydroxyphenyl)propane BPA

A10324-30 A10324-0B A10324-0I A10324-0I A10324-30 A10324-0B
AAA10324-0BEA 56.99 USD InStock
CAAAA10324-0I CAAAA10324-30 AAA10324-0B AAA10324-30 CAAAA10324-0B AAA10324-0I
4,4'-Isopropylidenediphenol 97+%
4,4'-Isopropylidenediphenol


Formula: C₁₅H₁₆O₂
MW: 228.29 g/mol
Boiling Pt: 200 °C/4 mm
Melting Pt: 153…158 °C
Density: 1.205 g/cm³ (20 °C)
Flash Pt: 227 °C(440 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002366
CAS Number: 80-05-7
EINECS: 201-245-8
UN: 3077
ADR: "9",III
Merck Index: 13,01296
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty