Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Arabinose 99%

Supplier: Thermo Scientific Chemicals
Synonyms: (2S,3R,4R)-2,3,4,5-Tetrahydroxypentanal

A10357-09 A10357-09 A10357-30 A10357-18 A10357-18 A10357-30
AAAA10357-09EA 45.69 USD InStock
AAAA10357-09 AAA10357-30 AAA10357-18 CAAAA10357-30 CAAAA10357-18 CAAAAA10357-09
D-(-)-Arabinose 99%
D(-)-Arabinose


Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Melting Pt: 154…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00135608
CAS Number: 10323-20-3
EINECS: 233-708-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty