Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(Methylthio)benzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A10379-14 A10379-06 A10379-06 A10379-14
AAA10379-06EA 109.99 USD InStock
CAAAA10379-14 AAA10379-06 CAAAA10379-06 AAA10379-14
2-(Methylthio)benzoic acid 98+%
2-(Methylthio)benzoic acid


Formula: C₈H₈O₂S
MW: 168.22 g/mol
Melting Pt: 166…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00029934
CAS Number: 3724-10-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty