Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzothiazole 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Benzo[d]thiazole

A10380-0B A10380-18 A10380-30 A10380-0B A10380-30 A10380-18
AAA10380-30EA 64.99 USD InStock
AAA10380-30 CAAAA10380-0B CAAAA10380-30 AAA10380-18 AAA10380-0B CAAAA10380-18
Benzothiazole 97%
Benzothiazole


Formula: C₇H₅NS
MW: 135.19 g/mol
Boiling Pt: 230 °C
Melting Pt: 2 °C
Density: 1.241
Flash Pt: 107 °C(224 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005775
CAS Number: 95-16-9
EINECS: 202-396-2
UN: 2810
ADR: "6.1",III
Merck Index: 13,01108
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty