Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Pyridylmethanol 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Pyridylmethanol Pyridine-3-methanol 3-Pyridylcarbinol

A10381-0B A10381-0B A10381-18 A10381-30 A10381-30 A10381-18
AAA10381-30EA 186.62 USD InStock
CAAAA10381-0B AAA10381-30 CAAAA10381-30 AAA10381-18 AAA10381-0B CAAAA10381-18
3-Pyridylmethanol 98%
3-Pyridylmethanol


Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 154 °C/28 mm
Melting Pt: –8 to –6 °C
Density: 1,130
Flash Pt: 149 °C(300 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00006407
CAS Number: 100-55-0
EINECS: 202-864-6
Merck Index: 12,06614
Order Now

 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty