Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Ethylanisole 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Ethyl-4-methoxybenzene

A10391-09 A10391-18 A10391-09 A10391-18
AAA10391-18EA 196.99 USD InStock
AAA10391-18 AAA10391-09 CAAAA10391-18 CAAAA10391-09
4-Ethylanisole 98+%
4-Ethylanisole


Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 195 °C
Density: 0.962
Flash Pt: 79 °C(174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015176
CAS Number: 1515-95-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty