Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Pentyn-1-ol 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Pentyn-5-ol

A10405-14 A10405-06 A10405-06 A10405-14 A10405-22 A10405-22
AAA10405-22EA 676.99 USD InStock
AAA10405-22 AAA10405-14 CAAAA10405-14 CAAAA10405-06 CAAAA10405-22 AAA10405-06
4-Pentyn-1-ol 97%
4-Pentyn-1-ol


Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 153…155 °C
Density: 0,907
Flash Pt: 62 °C(143 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002974
CAS Number: 5390-04-5
EINECS: 226-383-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty