Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,4-Dioxaspiro[4.5]decan-8-one 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1,4-Cyclohexanedione monoethylene ketal1,4-Cyclohexanedione monoethylene acetal

A10408-22 A10408-14 A10408-22 A10408-06 A10408-14 A10408-06
AAA10408-06EA 47.79 USD InStock
AAA10408-06 AAA10408-14 CAAAA10408-06 AAA10408-22 CAAAA10408-14 CAAAA10408-22
1,4-Dioxaspiro[4.5]decan-8-one 97%
1,4-Dioxaspiro[4.5]decan-8-one


Formula: C₈H₁₂O₃
MW: 156.18 g/mol
Melting Pt: 72…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00010214
CAS Number: 4746-97-8
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty