Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Azobenzene 97+%

Supplier: Thermo Scientific Chemicals
Danger

A10425-14 A10425-22 A10425-14 A10425-22 A10425-36 A10425-36
AAA10425-22EA 144.05 USD InStock
AAA10425-22 AAA10425-14 AAA10425-36 CAAAA10425-14 CAAAA10425-36 CAAAA10425-22
Azobenzene 97+%
Azobenzene


Formula: C₁₂H₁₀N₂
MW: 182.22 g/mol
Boiling Pt: 293 °C
Melting Pt: 65…69 °C
Density: 1.203
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00003022
CAS Number: 103-33-3
EINECS: 203-102-5
UN: 3077
ADR: "9",III
Merck Index: 12,00948
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty