Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromopropane 99% ACS

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: n-Propyl bromide Solvent 38

A10461-AU A10461-AK A10461-0J A10461-0J A10461-AK A10461-AU
AAA10461-AUEA 130.7 USD InStock
AAA10461-AU CAAAA10461-AK CAAAA10461-0J CAAAA10461-AU AAA10461-0J AAA10461-AK
1-Bromopropane 99% ACS
1-Bromopropane


Formula: BrH₂CCH₂CH₃
MW: 122.99 g/mol
Boiling Pt: 70…71 °C
Melting Pt: –110 °C
Density: 1.353
Flash Pt: 21 °C(70 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000254
CAS Number: 106-94-5
EINECS: 203-445-0
UN: 2344
ADR: "3",III
Merck Index: 12,08030
https://us-legacy.vwr.com/stibo/web/std.lang.all/75/23/717523.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty