Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α,α,α-Trifluoro-3-tolualdehyde 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-(Trifluoromethyl)benzaldehyde

A10463-22 A10463-22 A10463-14 A10463-06 A10463-06 A10463-14
AAA10463-22EA 592.99 USD InStock
CAAAA10463-22 AAA10463-22 AAA10463-14 AAA10463-06 CAAAA10463-06 CAAAA10463-14
α,α,α-Trifluoro-3-tolualdehyde 97%
α,α,α-Trifluoro-3-tolualdehyde


Formula: C₈H₅F₃O
MW: 174.12 g/mol
Boiling Pt: 83…86 °C/30 mm
Density: 1,304
Flash Pt: 68 °C(154 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003373
CAS Number: 454-89-7
EINECS: 207-228-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty