Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2-Naphthoquinone-4-sulfonic acid sodium salt 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Sodium 3,4-dioxo-3,4-dihydronaphthalene-1-sulfonateFolin's reagentSodium 3,4-dioxo-3,4-dihydronaphthalene-1-sulphonate

A10507-22 A10507-22 A10507-14 A10507-14
AAA10507-22EA 242.99 USD InStock
CAAAA10507-22 CAAAA10507-14 AAA10507-22 AAA10507-14
1,2-Naphthoquinone-4-sulfonic acid sodium salt 97%
1,2-Naphthoquinone-4-sulphonic acid sodium salt
Colorimetric determination of amino acids and amines.

Soluble in water. Moderately soluble in acetone. Slightly soluble in alcohol


Formula: C₁₀H₅NaO₅S
MW: 260.20 g/mol
Melting Pt: ca 289 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00001700
CAS Number: 521-24-4
EINECS: 208-308-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty