Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Valerophenone 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 1-Phenyl-1-pentanone

A10525-22 A10525-14 A10525-36 A10525-14 A10525-36 A10525-22
AAA10525-22EA 134.95 USD InStock
CAAAA10525-36 CAAAA10525-14 AAA10525-22 CAAAA10525-22 AAA10525-14 AAA10525-36
Valerophenone 99%
Valerophenone


Formula: C₆H₅C(O)(CH₂)₃CH₃
MW: 162.23 g/mol
Boiling Pt: 244…245 °C
Melting Pt: –9 °C
Density: 0.978
Flash Pt: 102 °C(215 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009480
CAS Number: 1009-14-9
EINECS: 213-767-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/22/93/762293.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty